Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174472.4 | NP_776897.2 | 3 - 4 | 642 | 100 | |
GenBank mRNA | AB099088.1 | - | 1 - 2 | 77 | 100 | |
AF144764.1 | - | 3 - 4 | 280 | 100 | ||
BC102710.1 | - | 3 - 4 | 644 | 100 | ||
M32303.1 | - | 3 - 4 | 631 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |