Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175824.2 | NP_787018.1 | 2 - 3 | 252 | 145 | |
GenBank mRNA | BC102330.1 | - | 2 - 3 | 252 | 145 | |
M33141.1 | - | 1 - 2 | 56 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |