Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175824.2 | NP_787018.1 | 4 - 5 | 379 | 107 | |
GenBank mRNA | BC102330.1 | - | 4 - 5 | 379 | 107 | |
M33141.1 | - | 3 - 4 | 183 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |