Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203362.2 | NP_976238.2 | 2 - 3 | 449 | 77 | |
GenBank mRNA | BC102413.1 | - | 2 - 3 | 449 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |