Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206843.2 | NP_996674.1 | 2 - 3 | 333 | 73 | |
GenBank mRNA | BC102291.1 | - | 2 - 3 | 333 | 73 | |
X99692.1 | - | 160 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |