Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001172.3 | NP_001001172.2 | 1 - 2 | 210 | 97 | |
GenBank mRNA | BC112574.1 | - | 1 - 2 | 211 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |