Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024509.3 | NP_001019680.2 | 2 - 3 | 183 | 117 | |
GenBank mRNA | BC105379.1 | - | 2 - 3 | 184 | 117 | |
DQ010410.1 | - | 2 - 3 | 101 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |