Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024549.2 | NP_001019720.1 | 4 - 5 | 1326 | 63 | |
GenBank mRNA | BC111208.1 | - | 4 - 5 | 1326 | 63 | |
BT021890.1 | - | 3 - 4 | 1218 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |