Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037595.2 | NP_001032684.1 | 1 - 2 | 139 | 80 | |
GenBank mRNA | BC109995.1 | - | 1 - 2 | 140 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |