Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037595.2 | NP_001032684.1 | 5 - 6 | 646 | 134 | |
GenBank mRNA | BC109995.1 | - | 5 - 6 | 647 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |