Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037610.2 | NP_001032699.1 | 1 - 2 | 260 | 99 | |
GenBank mRNA | BC109622.1 | - | 1 - 2 | 261 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |