Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037816.1 | NP_001032905.1 | 7 - 8 | 1017 | 104 | |
GenBank mRNA | BC111139.1 | - | 7 - 8 | 1017 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |