Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038037.2 | NP_001033126.1 | 2 - 3 | 201 | 115 | |
GenBank mRNA | BC109973.1 | - | 2 - 3 | 202 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |