Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038044.2 | NP_001033133.1 | 2 - 3 | 452 | 111 | |
GenBank mRNA | BC111283.1 | - | 2 - 3 | 453 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |