Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038048.2 | NP_001033137.1 | 3 - 4 | 588 | 99 | |
GenBank mRNA | BC109804.1 | - | 3 - 4 | 589 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |