Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038050.2 | NP_001033139.1 | 6 - 7 | 681 | 103 | |
GenBank mRNA | BC111178.1 | - | 6 - 7 | 682 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |