Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038076.2 | NP_001033165.2 | 3 - 4 | 421 | 79 | |
GenBank mRNA | BC111159.1 | - | 3 - 4 | 420 | 79 | |
EV643159.1 | - | 3 - 4 | 424 | 79 | ||
EV658551.1 | - | 3 - 4 | 425 | 79 | ||
EV746330.1 | - | 1 - 2 | 461 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |