Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038084.2 | NP_001033173.1 | 1 - 2 | 156 | 101 | |
GenBank mRNA | BC109828.1 | - | 1 - 2 | 157 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |