Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191293.2 | NP_001178222.2 | 1 - 2 | 177 | 105 | |
GenBank mRNA | BC109818.1 | - | 1 - 2 | 139 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |