Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038090.2 | NP_001033179.1 | 3 - 4 | 496 | 96 | |
GenBank mRNA | BC109917.1 | - | 3 - 4 | 497 | 96 | |
BC148921.1 | - | 2 - 3 | 1045 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |