Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038104.2 | NP_001033193.1 | 2 - 3 | 217 | 86 | |
GenBank mRNA | BC111244.1 | - | 2 - 3 | 218 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |