Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038114.2 | NP_001033203.1 | 3 - 4 | 562 | 110 | |
GenBank mRNA | BC109876.1 | - | 3 - 4 | 564 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |