Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038120.1 | NP_001033209.1 | 5 - 6 | 804 | 92 | |
GenBank mRNA | BC111229.1 | - | 5 - 6 | 804 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |