Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFB2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038127.2 | NP_001033216.1 | 4 - 5 | 863 | 121 | |
GenBank mRNA | BC109634.1 | - | 4 - 5 | 864 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |