Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C27H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038136.2 | NP_001033225.1 | 7 - 8 | 1271 | 121 | |
GenBank mRNA | BC111359.1 | - | 7 - 8 | 1272 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |