Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038162.2 | NP_001033251.1 | 1 - 2 | 152 | 121 | |
GenBank mRNA | BC111143.1 | - | 1 - 2 | 154 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |