Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGFG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038171.2 | NP_001033260.1 | 4 - 5 | 766 | 63 | |
GenBank mRNA | BC111120.1 | - | 4 - 5 | 767 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |