Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038184.1 | NP_001033273.1 | 10 - 11 | 1500 | 77 | |
GenBank mRNA | BC111331.1 | - | 10 - 11 | 1500 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |