Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038186.2 | NP_001033275.1 | 7 - 8 | 950 | 169 | |
GenBank mRNA | BC111158.1 | - | 7 - 8 | 951 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |