Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038199.2 | NP_001033288.1 | 2 - 3 | 339 | 81 | |
GenBank mRNA | BC109993.1 | - | 2 - 3 | 340 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |