Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038201.2 | NP_001033290.1 | 6 - 7 | 593 | 78 | |
GenBank mRNA | BC111270.1 | - | 6 - 7 | 596 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |