Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038218.2 | NP_001033307.1 | 10 - 11 | 1005 | 63 | |
GenBank mRNA | BC111188.1 | - | 10 - 11 | 1006 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |