Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038219.2 | NP_001033308.2 | 11 - 12 | 1661 | 106 | |
GenBank mRNA | BC109498.1 | - | 11 - 12 | 1662 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |