Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038221.2 | NP_001033310.1 | 18 - 19 | 2190 | 129 | |
GenBank mRNA | BC111193.1 | - | 18 - 19 | 2191 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |