Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038223.2 | NP_001033312.1 | 4 - 5 | 352 | 88 | |
GenBank mRNA | BC109806.1 | - | 4 - 5 | 353 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |