Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038509.2 | NP_001033598.1 | 6 - 7 | 726 | 80 | |
GenBank mRNA | BC110005.1 | - | 6 - 7 | 728 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |