Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038510.3 | NP_001033599.2 | 4 - 5 | 482 | 60 | |
GenBank mRNA | BC109524.1 | - | 4 - 5 | 483 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |