Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038510.3 | NP_001033599.2 | 7 - 8 | 1006 | 85 | |
GenBank mRNA | BC109524.1 | - | 7 - 8 | 1007 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |