Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038515.1 | NP_001033604.1 | 2 - 3 | 180 | 58 | |
GenBank mRNA | BC109888.1 | - | 2 - 3 | 180 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |