Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038520.2 | NP_001033609.1 | 5 - 6 | 476 | 61 | |
GenBank mRNA | BC109885.1 | - | 5 - 6 | 478 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |