Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038543.2 | NP_001033632.1 | 3 - 4 | 248 | 89 | |
GenBank mRNA | BC109905.1 | - | 3 - 4 | 249 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |