Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038545.2 | NP_001033634.1 | 1 - 2 | 66 | 90 | |
GenBank mRNA | BC109977.1 | - | 1 - 2 | 67 | 90 | |
BT030697.1 | - | 1 - 2 | 36 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |