Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SET | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038564.1 | NP_001033653.1 | 5 - 6 | 514 | 90 | |
GenBank mRNA | BC109772.1 | - | 5 - 6 | 514 | 90 | |
BT030717.1 | - | 6 - 7 | 860 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |