Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038620.1 | NP_001033709.1 | 1 - 2 | 185 | 152 | |
GenBank mRNA | AY923833.1 | - | 1 - 2 | 185 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |