Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038678.2 | NP_001033767.1 | 4 - 5 | 678 | 81 | |
GenBank mRNA | BC110171.1 | - | 4 - 5 | 679 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |