Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038678.2 | NP_001033767.1 | 7 - 8 | 961 | 76 | |
GenBank mRNA | BC110171.1 | - | 7 - 8 | 962 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |