Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038688.2 | NP_001033777.1 | 3 - 4 | 362 | 62 | |
GenBank mRNA | BC110229.1 | - | 3 - 4 | 363 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |