Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040529.2 | NP_001035619.1 | 5 - 6 | 519 | 102 | |
GenBank mRNA | BC110184.1 | - | 5 - 6 | 520 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |