Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040529.2 | NP_001035619.1 | 6 - 7 | 664 | 142 | |
GenBank mRNA | BC110184.1 | - | 6 - 7 | 665 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |