Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040529.2 | NP_001035619.1 | 8 - 9 | 896 | 71 | |
GenBank mRNA | BC110184.1 | - | 8 - 9 | 897 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |